missing translation for 'onlineSavingsMsg'
Learn More

4-Formylbenzenesulfonic Acid, TRC

Product Code. 30395806
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395806 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395806 Supplier Toronto Research Chemicals Supplier No. TRCF7547251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Formylbenzenesulfonic Acid
CAS 5363-54-2
Molecular Formula C7 H6 O4 S
Quantity 1 g
Synonym 4-Formylbenzenesulfonic Acid, Benzaldehyde-4-sulfonic Acid, p-Formylbenzenesulfonic Acid
InChI Formula InChI=1S/C7H6O4S/c8-5-6-1-3-7(4-2-6)12(9,10)11/h1-5H,(H,9,10,11)
SMILES OS(=O)(=O)c1ccc(C=O)cc1
IUPAC Name 4-formylbenzenesulfonic acid
Molecular Weight (g/mol) 186.185
Formula Weight 185.999
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.