missing translation for 'onlineSavingsMsg'
Learn More

4-Formylbenzeneboronic acid, 97%

Product Code. 11410678
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410678 1 g 1g
11420678 5 g 5g
11430678 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410678 Supplier Thermo Scientific Alfa Aesar Supplier No. B25199.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87199-17-5 | C7H7BO3 | 149.94 g/mol

4-Formylbenzeneboronic acid acts as a reagent used for Palladium-catalyzed arylation Suzuki-Miyaura cross-coupling in water, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, ligand-free copper-catalyzed coupling of nitro arenes with arylboronic acids, triethylamine-catalyzed three-component Hantzsch condensations, copper-catalyzed nitrations, oxidative mono-cleavage of dialkenes catalyzed by Trametes hirsuta, palladacycle-catalyzed cross-coupling of arylboronic acids with carboxylic anhydrides or acyl chlorides, palladium-catalyzed aerobic oxidative cross-coupling reactions. It acts as a reagent used in preparation of sensitizers with dithiafulvenyl unit as electron donor for high-efficiency dye-sensitized solar cells, a novel protein synthesis inhibitor active against Gram-positive bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Formylbenzeneboronic acid acts as a reagent used for Palladium-catalyzed arylation Suzuki-Miyaura cross-coupling in water, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, ligand-free copper-catalyzed coupling of nitro arenes with arylboronic acids, triethylamine-catalyzed three-component Hantzsch condensations, copper-catalyzed nitrations, oxidative mono-cleavage of dialkenes catalyzed by Trametes hirsuta, palladacycle-catalyzed cross-coupling of arylboronic acids with carboxylic anhydrides or acyl chlorides, palladium-catalyzed aerobic oxidative cross-coupling reactions. It acts as a reagent used in preparation of sensitizers with dithiafulvenyl unit as electron donor for high-efficiency dye-sensitized solar cells, a novel protein synthesis inhibitor active against Gram-positive bacteria.

Solubility
Slightly Soluble in water.

Notes
Air Sensitive. Store away from air and oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87199-17-5
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00151823
InChI Key VXWBQOJISHAKKM-UHFFFAOYSA-N
Synonym 4-formylphenyl boronic acid, 4-formylbenzeneboronic acid, 4-boronobenzaldehyde, boronic acid, 4-formylphenyl, boronic acid, b-4-formylphenyl, p-formylphenylboronic acid, 4-dihydroxyboryl benzaldehyde, 4-dihydroxyboranyl benzaldehyde, 4-benzaldehyde boronic acid, 4-formyl-phenylboronic acid
PubChem CID 591073
IUPAC Name (4-formylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=O)C=C1

Specifications

Melting Point 260°C to 266°C
Quantity 1 g
Beilstein 3030770
Sensitivity Air Sensitive
Solubility Information Slightly Soluble in water.
Formula Weight 149.94
Percent Purity 97%
Chemical Name or Material 4-Formylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.