missing translation for 'onlineSavingsMsg'
Learn More

4-Formyl-n-isopropylbenzamide, TRC

Product Code. 30394710
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394710 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394710 Supplier Toronto Research Chemicals Supplier No. TRCF7004255G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Formyl-n-isopropylbenzamide
CAS 13255-50-0
Molecular Formula C11H13NO2
Quantity 5 g
Synonym Benzamide, 4-formyl-N-(1-methylethyl)-, Terephthalaldehydamide, N-isopropyl- (8CI), 4-Formyl-N-(1-methylethyl)benzamide, 4-Formyl-N-isopropylbenzamide, N-Isopropyl-4-formylbenzamide, N-Isopropyl-p-formylbenzamide, N-Isopropylterephthalaldehydamide
InChI Formula InChI=1S/C11H13NO2/c1-8(2)12-11(14)10-5-3-9(7-13)4-6-10/h3-8H,1-2H3,(H,12,14)
SMILES CC(C)NC(=O)c1ccc(C=O)cc1
IUPAC Name 4-formyl-N-propan-2-ylbenzamide
Molecular Weight (g/mol) 191.23
Formula Weight 191.0946
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.