missing translation for 'onlineSavingsMsg'
Learn More

(4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl Dihydrogen Phosphate Hydrate, TRC

Product Code. 30010784
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010784 10 g 10g
30003237 5 g 5g
17998384 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010784 Supplier Toronto Research Chemicals Supplier No. TRCF70052310G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41468-25-1
Molecular Formula C8 H10 N O6 P . H2 O
Molecular Weight (g/mol) 265.16
Synonym 4-Pyridinecarboxaldehyde, 3-hydroxy-2-methyl-5-[(phosphonooxy)methyl]-, hydrate (1:1), 4-Pyridinecarboxaldehyde, 3-hydroxy-2-methyl-5-[(phosphonooxy)methyl]-, monohydrate (9CI), MC 1, Pyridoxal 5'-phosphate monohydrate
IUPAC Name (4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate;hydrate
SMILES O.Cc1ncc(COP(=O)(O)O)c(C=O)c1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H10NO6P.H2O/c1-5-8(11)7(3-10)6(2-9-5)4-15-16(12,13)14;/h2-3,11H,4H2,1H3,(H2,12,13,14);1H2
Formula Weight 265.0351
Chemical Name or Material Pyridoxal-5'-phosphate monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.