missing translation for 'onlineSavingsMsg'
Learn More

(4-Formyl-3-methoxyphenyl)boronic Acid, TRC

Product Code. 30402276
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402276 1 g
30389431 250 mg
30402386 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402276 Supplier Toronto Research Chemicals Supplier No. TRCF7547101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 815620-00-9
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.97
SMILES COc1cc(ccc1C=O)B(O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H9BO4/c1-13-8-4-7(9(11)12)3-2-6(8)5-10/h2-5,11-12H,1H3
Formula Weight 180.0594
Chemical Name or Material (4-Formyl-3-methoxyphenyl)boronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.