missing translation for 'onlineSavingsMsg'
Learn More

4-Formyl-2-methylresorcinol, TRC

Product Code. 30395501
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395501 1 g
30392620 10 g
30401536 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395501 Supplier Toronto Research Chemicals Supplier No. TRCF7008451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6248-20-0
Molecular Formula C8 H8 O3
Molecular Weight (g/mol) 152.15
Synonym β-Resorcylaldehyde, 3-methyl- (8CI), 2,4-Dihydroxy-3-methylbenzaldehyde, 2,4-Dihydroxy-m-tolualdehyde, 3-Methyl-2,4-dihydroxybenzaldehyde, 4-Formyl-2-methylresorcinol
IUPAC Name 2,4-dihydroxy-3-methylbenzaldehyde
SMILES Cc1c(O)ccc(C=O)c1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H8O3/c1-5-7(10)3-2-6(4-9)8(5)11/h2-4,10-11H,1H3
Formula Weight 152.0473
Chemical Name or Material 2,4-Dihydroxy-3-methylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.