missing translation for 'onlineSavingsMsg'
Learn More

4'-Formyl(1,1'-biphenyl)-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-3075638
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10025222

Brand: Thermo Scientific Maybridge MO01260DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 70916-98-2
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD03424613
InChI Key JCEAFZUEUSBESW-UHFFFAOYSA-N
Synonym 4-4-formylphenyl benzoic acid, 4'-formylbiphenyl-4-carboxylic acid, 4'-formyl-1,1'-biphenyl-4-carboxylic acid, 4'-formyl-biphenyl-4-carboxylic acid, 4-biphenyl-4'-formyl-carboxylic acid, 4'-formyl 1,1'-biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, 4'-formyl, 4-biphenyl-4'-formyl-carboxylicacid, pubchem10282
PubChem CID 2794722
SMILES OC(=O)C1=CC=C(C=C1)C1=CC=C(C=O)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 4-(4-formylphenyl)benzoic acid
Formula Weight 226.23
Percent Purity 97%
Chemical Name or Material 4'-Formyl(1, 1'-biphenyl)-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.