missing translation for 'onlineSavingsMsg'
Learn More

4-Flurobenzenesulfinic acid sodium salt, TRC

Product Code. 30392243
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392243 2.5 g 2.5g
30396472 500 mg -
30390569 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392243 Supplier Toronto Research Chemicals Supplier No. TRCF6006102.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-80-6
Molecular Formula C6H4FNaO2S
Molecular Weight (g/mol) 182.15
Synonym Benzenesulfinic acid, 4-fluoro-, sodium salt (1:1) (ACI), Benzenesulfinic acid, 4-fluoro-, sodium salt (9CI), Benzenesulfinic acid, p-fluoro-, sodium salt (6CI, 7CI, 8CI), 4-Fluorobenzenesulfinic acid sodium salt, NSC 131873, Sodium 4-fluorobenzenesulfinate, Sodium p-fluorobenzenesulfinate
IUPAC Name sodium;4-fluorobenzenesulfinate
SMILES [Na+].[O-]S(=O)c1ccc(F)cc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C6H5FO2S.Na/c7-5-1-3-6(4-2-5)10(8)9;/h1-4H,(H,8,9);/q;+1/p-1
Formula Weight 181.9814
Chemical Name or Material 4-Flurobenzenesulfinic acid sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.