missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorothiophenol, 97%

Product Code. 15730799 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15730799 50 g Glass Bottle 50g
10580591 10 g Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15730799 Supplier Thermo Scientific Acros Supplier No. 119650500
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 371-42-6 | C6H5FS | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-42-6
Molecular Formula C6H5FS
Molecular Weight (g/mol) 128.17
MDL Number MFCD00004846
InChI Key OKIHXNKYYGUVTE-UHFFFAOYSA-N
Synonym 4-fluorothiophenol, p-fluorothiophenol, 4-fluorobenzene-1-thiol, 4-fluoro thiophenol, p-fluorophenyl mercaptan, benzenethiol, 4-fluoro, 4-fluoro-benzenethiol, 4-mercaptofluorobenzene, 4-fluorthiophenol, p-fluoro-thiophenol
PubChem CID 67789
IUPAC Name 4-fluorobenzenethiol
SMILES C1=CC(=CC=C1F)S

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 54g/mL
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula FC6H4SH
Refractive Index 1.5480 to 1.5500
Quantity 50 g
Formula Weight 128.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Fluorothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.