missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorostyrene, 97+%, stab. with 0.1% 4-tert-butylcatechol

Product Code. 11472487
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472487 5 g 5g
11374719 1 g 1g
11482487 25 g 25g
11492487 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11472487 Supplier Thermo Scientific Alfa Aesar Supplier No. A14775.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 405-99-2 | C8H7F | 122.142 g/mol

4-Fluorostyrene, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorostyrene, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from acids, bases, oxidizing agents, halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 405-99-2
Molecular Formula C8H7F
Molecular Weight (g/mol) 122.142
MDL Number MFCD00000361
InChI Key JWVTWJNGILGLAT-UHFFFAOYSA-N
Synonym 4-fluorostyrene, 1-fluoro-4-vinylbenzene, p-fluorostyrene, benzene, 1-ethenyl-4-fluoro, styrene, p-fluoro, para-fluorostyrene, 1-fluoro-4-vinyl-benzene, 4-fluoro-styrene, acmc-1cszp
PubChem CID 67883
IUPAC Name 1-ethenyl-4-fluorobenzene
SMILES C=CC1=CC=C(C=C1)F

Specifications

Melting Point -36°C
Density 1.024
Boiling Point 145°C to 148°C
Flash Point 26°C (78°F)
Odor Recognizable
Refractive Index 1.515
Quantity 5 g
UN Number UN1993
Beilstein 1634203
Solubility Information Insoluble in water.
Formula Weight 122.14
Percent Purity ≥97%
Chemical Name or Material 4-Fluorostyrene, stabilized with 0.1% 4-tert-butylcatechol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.