missing translation for 'onlineSavingsMsg'
Learn More

4-Fluororesorcinol, 97%

Product Code. 15486888
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486888 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15486888 Supplier Thermo Scientific Alfa Aesar Supplier No. H60426.ME
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103068-41-3 | C6H5FO2 | 128.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 103068-41-3
Melting Point 96°C to 100°C
Molecular Formula C6H5FO2
MDL Number MFCD03789074
Quantity 500 mg
Synonym 4-fluororesorcinol, 4-fluoro-1,3-benzenediol, 1-fluoro-2,4-dihydroxybenzene, 4-fluoro-1,3-dihydroxybenzene, 1,3-benzenediol, 4-fluoro, 2,4-dihydroxyfluorobenzene, 4fluororesorcinol, pubchem22656, 4-fluoroben-zene-1,3-diol
InChI Key XPOIJNIQXJYQOV-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1O)O)F
IUPAC Name 4-fluorobenzene-1,3-diol
Molecular Weight (g/mol) 128.102
PubChem CID 10558640
Formula Weight 128.1
Percent Purity 97%
Chemical Name or Material 4-Fluororesorcinol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluororesorcinol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Fluororesorcinol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.