missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophthalic anhydride, 98%

Product Code. 11459287
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459287 25 g 25g
11449287 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459287 Supplier Thermo Scientific Alfa Aesar Supplier No. B21352.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 319-03-9 | C8H3FO3 | 166.11 g/mol

4-Fluorophthalic anhydride is used in the preparation of 5-fluoro-2-pyridin-3-ylmethyl-isoindole-1,3-dione by reacting with C-pyridin-3-yl-methylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophthalic anhydride is used in the preparation of 5-fluoro-2-pyridin-3-ylmethyl-isoindole-1,3-dione by reacting with C-pyridin-3-yl-methylamine.

Solubility
Soluble in sodium hydroxide.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 319-03-9
Molecular Formula C8H3FO3
Molecular Weight (g/mol) 166.11
MDL Number MFCD00191363
InChI Key XVMKZAAFVWXIII-UHFFFAOYSA-N
Synonym 4-fluorophthalic anhydride, 5-fluoroisobenzofuran-1,3-dione, 5-fluoro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 5-fluoro, 4-fluorophthalicanhydride, 4-fluorophthalic acid anhydride, 5-fluoro-1,3-dihydro-2-benzofuran-1,3-dione, 4-fluorobenzene-1,2-dicarboxylic anhydride, 5-fluoro-isobenzofuran-1,3-dione
PubChem CID 67572
IUPAC Name 5-fluoro-2-benzofuran-1,3-dione
SMILES FC1=CC=C2C(=O)OC(=O)C2=C1

Specifications

Melting Point 74°C to 79°C
Density 1.5
Boiling Point 147°C to 149°C (20 mmHg)
Quantity 25 g
Beilstein 131281
Sensitivity Moisture sensitive
Solubility Information Soluble in sodium hydroxide.
Formula Weight 166.11
Percent Purity 98%
Chemical Name or Material 4-Fluorophthalic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.