missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophthalic acid, 98%

Product Code. 15494187
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494187 10 g 10g
15484187 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494187 Supplier Thermo Scientific Alfa Aesar Supplier No. H31597.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 320-97-8 | C8H5FO4 | 184.12 g/mol

4-Fluorophthalic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophthalic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 320-97-8
Molecular Formula C8H5FO4
Molecular Weight (g/mol) 184.12
MDL Number MFCD06208213
InChI Key OMCXTFVBNCFZMY-UHFFFAOYSA-N
Synonym 4-fluorobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-fluoro, 5-fluorophtalic acid, 5-fluorophthalic acid, 4-fluorobenzene-1,2-dioic acid, 3-fluorophthalicacid, pubchem1406, rarechem al bo 0829, 1,2-benzenedicarboxylicacid, 4-fluoro
PubChem CID 15312
IUPAC Name 4-fluorophthalic acid
SMILES OC(=O)C1=CC=C(F)C=C1C(O)=O

Specifications

Melting Point 154°C to 158°C
Odor Odorless
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 184.12
Percent Purity 98%
Chemical Name or Material 4-Fluorophthalic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.