missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylsulfonylacetonitrile, 97%

Product Code. 11428757
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428757 1 g 1g
11438757 5 g 5g
11448757 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428757 Supplier Thermo Scientific Alfa Aesar Supplier No. A19916.03
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 32083-66-2 | C8H6FNO2S | 199.199 g/mol

4-Fluorophenylsulfonylacetonitrile is used to produce 3-(4-fluoro-benzenesulfonyl)-thiophen-2-ylamine with [1,4]dithiane-2,5-diol. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylsulfonylacetonitrile is used to produce 3-(4-fluoro-benzenesulfonyl)-thiophen-2-ylamine with [1,4]dithiane-2,5-diol. It is also used as pharmaceutical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32083-66-2
Molecular Formula C8H6FNO2S
Molecular Weight (g/mol) 199.199
MDL Number MFCD00179339
InChI Key WBXJZTLPEMITJL-UHFFFAOYSA-N
Synonym 4-fluorobenzenesulfonyl acetonitrile, 2-4-fluorophenyl sulfonyl acetonitrile, 4-fluorobenzenesulphonyl acetonitrile, 4-fluorophenyl sulfonyl acetonitrile, 2-4-fluorobenzenesulfonyl acetonitrile, 4-fluorophenylsulfonylacetonitrile, 4-fluorobenzenesulfonylacetonitrile, 2-4-fluorophenyl sulfonylacetonitrile, 4-fluorophenylsulfonyl acetonitrile
PubChem CID 565732
IUPAC Name 2-(4-fluorophenyl)sulfonylacetonitrile
SMILES C1=CC(=CC=C1F)S(=O)(=O)CC#N

Specifications

Melting Point 89°C to 93°C
Quantity 1 g
UN Number UN3439
Formula Weight 199.2
Percent Purity 97%
Chemical Name or Material 4-Fluorophenylsulfonylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.