missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog Number 10240703
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
352-13-6, 109-99-9
C6H4BrFMg
199.31
MFCD00009667
QYBFFRXNNFXREA-UHFFFAOYSA-M
4-fluorophenylmagnesium bromide, magnesium, bromo 4-fluorophenyl, bromo 4-fluorophenyl magnesium, p-fluorophenylmagnesium bromide, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-fluorphenylmagnesium bromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide, grignard reagent
2734897
bromo(4-fluorophenyl)magnesium
FC1=CC=C([Mg]Br)C=C1
This item is not returnable. View return policy
352-13-6, 109-99-9
C6H4BrFMg
199.31
MFCD00009667
QYBFFRXNNFXREA-UHFFFAOYSA-M
4-fluorophenylmagnesium bromide, magnesium, bromo 4-fluorophenyl, bromo 4-fluorophenyl magnesium, p-fluorophenylmagnesium bromide, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-fluorphenylmagnesium bromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide, grignard reagent
2734897
bromo(4-fluorophenyl)magnesium
FC1=CC=C([Mg]Br)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Melting Point 5.0°C
Linear Formula F(C6H6)MgBr
Density 1.0100g/mL
Solubility Information Solubility in water: vigorous reaction.
Formula Weight 199.3
Percent Purity 14 to 17%
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.01
Quantity 100 mL
Flash Point −21°C
Physical Form Solution
Chemical Name or Material 4-Fluorophenylmagnesium bromide, 0.8M Solution in THF
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Fluorophenylmagnesium bromide, 0.8M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H261-In contact with water releases flammable gases.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'