missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylhydrazine hydrochloride, 97%

Product Code. 10195513 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10195513 100 g Glass bottle 100g
10570761 10 g Glass bottle 10g
10254770 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10195513 Supplier Thermo Scientific Acros Supplier No. 119591000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 823-85-8 | C6H8ClFN2 | 162.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 823-85-8
Molecular Formula C6H8ClFN2
Molecular Weight (g/mol) 162.59
MDL Number MFCD00012942
InChI Key FEKUXLUOKFSMRO-UHFFFAOYSA-N
Synonym 4-fluorophenylhydrazine hydrochloride, 4-fluorophenyl hydrazine hydrochloride, 4-fluorophenylhydrazine hcl, 1-4-fluorophenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine hcl, p-fluorophenylhydrazine hydrochloride, 4-fluorphenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine monohydrochloride, p-fluorophenylhydrazine hcl, hydrazine, 4-fluorophenyl-, monohydrochloride
PubChem CID 69981
SMILES [H+].[Cl-].NNC1=CC=C(F)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 275°C to 277°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula FC6H4NHNH2·HCl
Quantity 100 g
Beilstein 15, I, 105
Solubility Information Solubility in water: soluble
IUPAC Name (4-fluorophenyl)hydrazine;hydrochloride
Formula Weight 162.59
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Fluorophenylhydrazine hydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Fluorophenylhydrazine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.