missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetic acid, 98%

Product Code. 11433384
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433384 5 g 5g
11443384 25 g 25g
11453384 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433384 Supplier Thermo Scientific Alfa Aesar Supplier No. A10646.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 405-50-5 | C8H7FO2 | 154.14 g/mol

4-Fluorophenylacetic acid is used as an intermediate in the production of fluorinated anesthetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylacetic acid is used as an intermediate in the production of fluorinated anesthetics.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 405-50-5
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004343
InChI Key MGKPFALCNDRSQD-UHFFFAOYSA-N
Synonym 4-fluorophenylacetic acid, 2-4-fluorophenyl acetic acid, p-fluorophenylacetic acid, 4-fluorophenyl acetic acid, benzeneacetic acid, 4-fluoro, 4-fluorobenzeneacetic acid, p-fluorophenyl acetic acid, acetic acid, p-fluorophenyl, unii-s5yjv7caz1, 4-fluoro-phenyl-acetic acid
PubChem CID 9837
IUPAC Name 2-(4-fluorophenyl)acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)F

Specifications

Melting Point 81°C to 85°C
Boiling Point 164°C (2 mmHg)
Flash Point >100°C (212°F)
Odor Odorless
Quantity 5 g
Beilstein 972145
Merck Index 14,4177
Solubility Information Insoluble in water.
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 4-Fluorophenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Fluorophenylaceticacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.