missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl chloroformate, TRC

Product Code. 30399518
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399518 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399518 Supplier Toronto Research Chemicals Supplier No. TRCF1700001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Fluorophenyl chloroformate
CAS 38377-38-7
Molecular Formula C7 H4 Cl F O2
Quantity 1 g
Synonym Carbonochloridic acid, 4-fluorophenyl ester, p-Fluorophenyl chloroformate
InChI Formula InChI=1S/C7H4ClFO2/c8-7(10)11-6-3-1-5(9)2-4-6/h1-4H
SMILES Fc1ccc(OC(=O)Cl)cc1
IUPAC Name (4-fluorophenyl) carbonochloridate
Molecular Weight (g/mol) 174.557
Formula Weight 173.988
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.