missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoropentedrone Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Fluoropentedrone Hydrochloride |
| Analyte or Component Names | 4-Fluoropentedrone Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H16 F N O . Cl H |
| Synonym | 4-fluoro Pentedrone hydrochloride, 4-Fluoropentedrone hydrochloride, 4-F-Pentedrone hydrochloride, 1-(4-Fluorophenyl)-2-(methylamino)pentan-1-one hydrochloride, 4-Fluoro-α-methylaminopentiophenone hydrochloride, 4-Fluoro-α-methylaminovalerophenone hydrochloride, 4-FPD HCl |
| InChI Formula | InChI=1S/C12H16FNO.ClH/c1-3-4-11(14-2)12(15)9-5-7-10(13)8-6-9;/h5-8,11,14H,3-4H2,1-2H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CCCC(NC)C(=O)c1ccc(F)cc1 |
| IUPAC Name | 1-(4-fluorophenyl)-2-(methylamino)pentan-1-one;hydrochloride |
| Industry Type | Forensics |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?