missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoronaphthalene-1-boronic acid, 98%, Thermo Scientific™

Product Code. 15469447
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182344-25-8
Molecular Formula C10H8BFO2
Molecular Weight (g/mol) 189.98
MDL Number MFCD04039221
InChI Key IDCGAEJGZLYEAZ-UHFFFAOYSA-N
Synonym 4-fluoronaphthalene-1-boronic acid, 4-fluoronaphtalene-1-boronic acid, 4-fluoronaphthalen-1-yl-1-boronic acid, 4-fluoronaphthalen-1-yl boronic acid, 4-fluoro-1-naphthyl boronic acid, 4-fluoro-1-naphthaleneboronic acid, boronic acid, 4-fluoro-1-naphthalenyl, akos brn-0210, acmc-209ejb, 4-fluoronaphth-1-ylboronic acid
PubChem CID 2782675
IUPAC Name (4-fluoronaphthalen-1-yl)boronic acid
SMILES B(C1=CC=C(C2=CC=CC=C12)F)(O)O

Specifications

Melting Point 240°C to 242°C
Quantity 250 mg
Formula Weight 189.98
Percent Purity 98%
Chemical Name or Material 4-Fluoronaphthalene-1-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'