4-Fluoroisoindoline hydrochloride, 97%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 435860050

5GR 4-Fluoroisoindoline hydrochloride, 97%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 924305-06-6 Weight: 0.05500kg


Packaging Glass bottle
Quantity 5g

Product Code. 10565870

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification

Chemical Identifiers

Chemical Name or Material 4-Fluoroisoindoline hydrochloride
CAS 924305-06-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H8FN·ClH
Formula Weight 173.62

Specification

Physical Form Powder
Assay ≥96.0%
Formula Weight 173.62
Quantity 5g
Color White to off-white
CAS Min % 96.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 924305-06-6
Infrared Spectrum Authentic
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Water 5% Max. (K.F.)
Packaging Glass bottle
Molecular Formula C8H8FN·ClH
CAS Max % 100.0
Chemical Name or Material 4-Fluoroisoindoline hydrochloride
Alternative Products
Documentation

Technical documents

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.