missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorodiphenyl ether, 99%

Product Code. 11459327
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459327 100 g 100g
11439327 5 g 5g
11449327 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459327 Supplier Thermo Scientific Alfa Aesar Supplier No. B21380.22
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 330-84-7 | C12H9FO | 188.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330-84-7
Molecular Formula C12H9FO
Molecular Weight (g/mol) 188.20
MDL Number MFCD00055239
InChI Key AODSTUBSNYVSSL-UHFFFAOYSA-N
Synonym 4-fluorodiphenyl ether, 4-fluorophenyl phenyl ether, benzene, 1-fluoro-4-phenoxy, 4-fluoro-1-phenoxybenzene, 4-fluorodiphenylether, 4-fluorophenoxybenzene, pubchem1899, 4-fluoroduphenyl ether, 4-fluorophenylphenylether, #
PubChem CID 67614
IUPAC Name 1-fluoro-4-phenoxybenzene
SMILES FC1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Refractive Index 1.557
Quantity 100 g
Formula Weight 188.2
Percent Purity 99%
Chemical Name or Material 4-Fluorodiphenyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.