missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzylamine, 97%, Thermo Scientific Chemicals

Catalog Number 10265700
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
140-75-0
IIFVWLUQBAIPMJ-UHFFFAOYSA-N
4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392
67326
(4-fluorophenyl)methanamine
C1=CC(=CC=C1CN)F
This item is not returnable. View return policy
140-75-0
IIFVWLUQBAIPMJ-UHFFFAOYSA-N
4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392
67326
(4-fluorophenyl)methanamine
C1=CC(=CC=C1CN)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.09g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H8FN
Linear Formula FC6H4CH2NH2
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
Quantity 5 g
Molecular Weight (g/mol) 125.146
Formula Weight 125.15
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'