missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl mercaptan, 96%

Product Code. p-7080559
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11409382

Brand: Thermo Scientific Alfa Aesar A10126.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15894-04-9 | C7H7FS | 142.191 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15894-04-9
Molecular Formula C7H7FS
Molecular Weight (g/mol) 142.191
MDL Number MFCD00040724
InChI Key RKTRHMNWVZRZJQ-UHFFFAOYSA-N
Synonym 4-fluorobenzyl mercaptan, 4-fluorophenyl methanethiol, 4-fluorobenzylmercaptan, 4-fluoro benzyl mercaptan, 4-fluorobenzenemethanethiol, p-fluorotoluene-alpha-thiol, benzenemethanethiol, 4-fluoro, 4-fluoro-phenyl-methanethiol, 4-fluorobenzylthiol, 4-fluorobenzyl thiol
PubChem CID 85171
IUPAC Name (4-fluorophenyl)methanethiol
SMILES C1=CC(=CC=C1CS)F

Specifications

Density 1.157
Boiling Point 72°C to 74°C (15 mmHg)
Flash Point 76°C (168°F)
Refractive Index 1.543
Quantity 1 g
UN Number UN2810
Beilstein 2242520
Sensitivity Air Sensitive
Formula Weight 142.2
Percent Purity 96%
Chemical Name or Material 4-Fluorobenzyl mercaptan
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.