missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl Bromide, TRC

Product Code. 30399137
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30399137 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399137 Supplier Toronto Research Chemicals Supplier No. TRCF58861050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Fluorobenzyl Bromide
CAS 459-46-1
Molecular Formula C7H6BrF
Quantity 50 g
Synonym 1-(Bromomethyl)-4-fluorobenzene, p-Fluorobenzyl Bromide, alpha-Bromo-4-fluorotoluene, alpha-Bromo-p-fluorotoluene
InChI Formula InChI=1S/C7H6BrF/c8-5-6-1-3-7(9)4-2-6/h1-4H,5H2
SMILES Fc1ccc(CBr)cc1
IUPAC Name 1-(bromomethyl)-4-fluorobenzene
Molecular Weight (g/mol) 189.02
Formula Weight 187.9637
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.