missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl bromide, 97%

Product Code. 11439532
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439532 50 g 50g
11429532 10 g 10g
11449532 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439532 Supplier Thermo Scientific Alfa Aesar Supplier No. A10208.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 459-46-1 | C7H6BrF | 189.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-46-1
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.03
MDL Number MFCD00000359
InChI Key NVNPLEPBDPJYRZ-UHFFFAOYSA-N
Synonym 4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide
PubChem CID 68021
IUPAC Name 1-(bromomethyl)-4-fluorobenzene
SMILES FC1=CC=C(CBr)C=C1

Specifications

Density 1.53
Boiling Point 84°C to 85°C (15 mmHg)
Flash Point 66°C (150°F)
Odor Pungent
Refractive Index 1.547
Quantity 50 g
UN Number UN3265
Beilstein 636507
Formula Weight 189.03
Percent Purity 97%
Chemical Name or Material 4-Fluorobenzyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.