missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoyl chloride, 98%

Product Code. 10193580 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193580 100 mL Glass bottle 100mL
10677312 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10193580 Supplier Thermo Scientific Acros Supplier No. 152951000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 403-43-0 | C7H4ClFO | 158.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-43-0
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD00000684
InChI Key CZKLEJHVLCMVQR-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-fluoro, p-fluorobenzoyl chloride, benzoyl chloride, p-fluoro, p-fluorobenzoic acid chloride, 4-fluorobenzoylchloride, fluorobenzoyl 4-chloride, 4-fluorobenzoic acid chloride, p-fluorobenzoylchloride, p-fluorobenzoyl choride, 4-flurobenzoyl chloride
PubChem CID 67879
IUPAC Name 4-fluorobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 9.0°C
Color Colorless to Yellow
Density 1.3400g/mL
Boiling Point 82.0°C (20.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4COCl
Refractive Index 1.5299 to 1.5319
Quantity 100 mL
Beilstein 09, I, 137
Specific Gravity 1.34
Solubility Information Solubility in water: decomposes
Formula Weight 158.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzoyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.