missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzotrifluoride, 97%, Thermo Scientific™

Product Code. 10246630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246630 5 g Glass bottle 5g
10419690 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10246630 Supplier Thermo Scientific Acros Supplier No. 187390050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-44-8
Molecular Formula C7H4F4
Molecular Weight (g/mol) 164.103
MDL Number MFCD00000399
InChI Key UNNNAIWPDLRVRN-UHFFFAOYSA-N
Synonym 4-fluorobenzotrifluoride, 1-fluoro-4-trifluoromethyl benzene, p-fluorobenzotrifluoride, benzene, 1-fluoro-4-trifluoromethyl, alpha,alpha,alpha,4-tetrafluorotoluene, p-fluorotrifluoromethylbenzene, p-trifluoromethyl fluorobenzene, 4-flourobenzotrifluoride, a,a,a,4-tetrafluorotoluene, 4-trifluoromethyl fluorobenzene
PubChem CID 67873
IUPAC Name 1-fluoro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -41.7°C
Color Colorless
Density 1.2900g/mL
Boiling Point 102.0°C to 105.0°C
Flash Point <1°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula FC6H4CF3
Refractive Index 1.4004 to 1.4024
Quantity 5 g
Beilstein 05, III, 680
Formula Weight 164.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzotrifluoride, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Fluorobenzotrifluoride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.