missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzophenone, TRC

Product Code. 30404427
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404427 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404427 Supplier Toronto Research Chemicals Supplier No. TRCF588580100G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Fluorobenzophenone
CAS 345-83-5
Molecular Formula C13H9FO
Quantity 100 g
Synonym (4-Fluorophenyl)phenyl-methanone, 4-Fluoro-benzophenone, (4-Fluorophenyl)(phenyl)methanone, 4-Fluorophenyl Phenyl Ketone, NSC 88321, Phenyl 4-Fluorophenyl Ketone, p-Fluorobenzophenone, p-Fluorophenyl Phenyl Ketone
InChI Formula InChI=1S/C13H9FO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9H
SMILES Fc1ccc(cc1)C(=O)c2ccccc2
IUPAC Name (4-fluorophenyl)-phenylmethanone
Molecular Weight (g/mol) 200.21
Formula Weight 200.0637
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.