missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzophenone 98%, Thermo Scientific™

Product Code. 10493765 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10493765 10 g Glass bottle 10g
10071000 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10493765 Supplier Thermo Scientific Acros Supplier No. 119380100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 345-83-5
Molecular Formula C13H9FO
Molecular Weight (g/mol) 200.21
MDL Number MFCD00000352
InChI Key OGTSHGYHILFRHD-UHFFFAOYSA-N
Synonym 4-fluorobenzophenone, 4-fluorophenyl phenyl methanone, p-fluorobenzophenone, methanone, 4-fluorophenyl phenyl, 4-fluorophenyl phenyl ketone, 4-fluorophenyl-phenylmethanone, 4'-fluorobenzophenone, 4-fluoro-benzophenone, pubchem3308, benzophenone, 4-fluoro
PubChem CID 67663
IUPAC Name (4-fluorophenyl)(phenyl)methanone
SMILES FC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 43°C to 50°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H4COC6H5
Quantity 10 g
Beilstein 07, I, 226
Formula Weight 200.21
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzophenone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.