missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzophenone, 97%

Product Code. 11409133
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409133 100 g 100g
11499123 25 g 25g
11419133 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409133 Supplier Thermo Scientific Alfa Aesar Supplier No. A11185.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 345-83-5 | C13H9FO | 200.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345-83-5
Molecular Formula C13H9FO
Molecular Weight (g/mol) 200.21
MDL Number MFCD00000352
InChI Key OGTSHGYHILFRHD-UHFFFAOYSA-N
Synonym 4-fluorobenzophenone, 4-fluorophenyl phenyl methanone, p-fluorobenzophenone, methanone, 4-fluorophenyl phenyl, 4-fluorophenyl phenyl ketone, 4-fluorophenyl-phenylmethanone, 4'-fluorobenzophenone, 4-fluoro-benzophenone, pubchem3308, benzophenone, 4-fluoro
PubChem CID 67663
IUPAC Name (4-fluorophenyl)-phenylmethanone
SMILES FC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 45°C to 49°C
Boiling Point 130°C to 132°C (3 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 1869800
Formula Weight 200.21
Percent Purity 97%
Chemical Name or Material 4-Fluorobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.