missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzonitrile, TRC

Product Code. 30397031
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30397031 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30397031

Brand: Toronto Research Chemicals TRCF58837550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Fluorobenzonitrile
CAS 1194-02-1
Molecular Formula C7H4FN
Quantity 50 g
Synonym Benzonitrile, 4-fluoro-, Benzonitrile, p-fluoro- (6CI,7CI,8CI), 4-Fluorobenzonitrile, 1-Cyano-4-fluorobenzene, 4-Cyanofluorobenzene, 4-Fluorocyanobenzene, NSC 88330, Para-fluorobenzonitrile, p-Cyanofluorobenzene, p-Fluorobenzonitrile, p-Fluorophenyl cyanide
InChI Formula InChI=1S/C7H4FN/c8-7-3-1-6(5-9)2-4-7/h1-4H
SMILES Fc1ccc(cc1)C#N
IUPAC Name 4-fluorobenzonitrile
Molecular Weight (g/mol) 121.11
Formula Weight 121.0328
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.