missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoic anhydride, 97%

Product Code. p-7032165
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15456347

Brand: Thermo Scientific Alfa Aesar H33527.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25569-77-1 | C14H8F2O3 | 262.21 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25569-77-1
Molecular Formula C14H8F2O3
Molecular Weight (g/mol) 262.21
MDL Number MFCD00425938
InChI Key BBLXFRIGTQYGOT-UHFFFAOYSA-N
Synonym 4-fluorobenzoic anhydride, p-fluorobenzoic anhydride, benzoic acid, p-fluoro-, anhydride, benzoic acid, 4-fluoro-, anhydride, 4-fluorobenzoicanhydride, 4-fluorophenyl anhydride #, bis p-fluorobenzoic anhydride, 4-fluorobenzoic acid, anhydride, 4-fluorobenzoyl 4-fluorobenzoate, 4-fluorobenzene-1-carboxylic anhydride
PubChem CID 579438
IUPAC Name (4-fluorobenzoyl) 4-fluorobenzoate
SMILES FC1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(F)C=C1

Specifications

Melting Point 111°C to 115°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 262.21
Percent Purity 97%
Chemical Name or Material 4-Fluorobenzoic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.