missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoic anhydride, 97%

Product Code. 15456347
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456347 1 g 1g
15466347 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456347 Supplier Thermo Scientific Alfa Aesar Supplier No. H33527.03
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25569-77-1 | C14H8F2O3 | 262.21 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25569-77-1
Molecular Formula C14H8F2O3
Molecular Weight (g/mol) 262.21
MDL Number MFCD00425938
InChI Key BBLXFRIGTQYGOT-UHFFFAOYSA-N
Synonym 4-fluorobenzoic anhydride, p-fluorobenzoic anhydride, benzoic acid, p-fluoro-, anhydride, benzoic acid, 4-fluoro-, anhydride, 4-fluorobenzoicanhydride, 4-fluorophenyl anhydride #, bis p-fluorobenzoic anhydride, 4-fluorobenzoic acid, anhydride, 4-fluorobenzoyl 4-fluorobenzoate, 4-fluorobenzene-1-carboxylic anhydride
PubChem CID 579438
IUPAC Name (4-fluorobenzoyl) 4-fluorobenzoate
SMILES FC1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(F)C=C1

Specifications

Melting Point 111°C to 115°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 262.21
Percent Purity 97%
Chemical Name or Material 4-Fluorobenzoic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.