missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzenesulfonyl Chloride, TRC

Product Code. 30397861
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397861 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397861 Supplier Toronto Research Chemicals Supplier No. TRCF5883351KG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Fluorobenzenesulfonyl Chloride
CAS 349-88-2
Molecular Formula C6H4ClFO2S
Quantity 1 kg
Synonym 4-Fluoro-benzenesulfonyl Chloride, p-Fluoro-benzenesulfonyl Chloride, 4-Fluorobenzene-1-sulfonyl Chloride, 4-Fluorophenylsulfonyl Chloride, NSC 140128, p-Fluorobenzenesulfonyl Chloride, p-Fluorophenylsulfonyl Chloride
InChI Formula InChI=1S/C6H4ClFO2S/c7-11(9,10)6-3-1-5(8)2-4-6/h1-4H
SMILES Fc1ccc(cc1)S(=O)(=O)Cl
IUPAC Name 4-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol) 194.61
Formula Weight 193.9605
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.