missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzenesulfonamide, 98+%

Product Code. 15482198
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482198 1 g 1g
15402208 250 mg 250mg
15492198 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15482198 Supplier Thermo Scientific Alfa Aesar Supplier No. H55834.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 402-46-0 | C6H6FNO2S | 175.177 g/mol

4-Fluorobenzenesulfonamide, is used to produce bis-(4-fluoro-benzenesulfonyl)-amine with 4-fluoro-benzenesulfonyl chloride at temperature of 55 - 60. This reaction will need reagent OH-.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzenesulfonamide, is used to produce bis-(4-fluoro-benzenesulfonyl)-amine with 4-fluoro-benzenesulfonyl chloride at temperature of 55 - 60. This reaction will need reagent OH-.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-46-0
Molecular Formula C6H6FNO2S
Molecular Weight (g/mol) 175.177
MDL Number MFCD00025384
InChI Key LFLSATHZMYYIAQ-UHFFFAOYSA-N
Synonym p-fluorobenzenesulfonamide, 4-fluorobenzenesulphonamide, 4-fluorobenzene-1-sulfonamide, p-fluorobenzenesulphonamide, 4-flourobenzenesulfonamide, 4-fluoro-benzenesulfonamide, benzenesulfonamide, p-fluoro, benzenesulfonamide, 4-fluoro, pubchem2167, acmc-1acsj
PubChem CID 120231
IUPAC Name 4-fluorobenzenesulfonamide
SMILES C1=CC(=CC=C1F)S(=O)(=O)N

Specifications

Melting Point 124°C to 127°C
Quantity 1 g
UN Number UN2811
Solubility Information It is soluble in water.
Formula Weight 175.18
Percent Purity ≥98%
Chemical Name or Material 4-Fluorobenzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.