missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzamidoxime, 98%, Thermo Scientific™

Product Code. 10217767 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10217767 1 g Glass bottle 1g
10720550 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10217767 Supplier Thermo Scientific Acros Supplier No. 429950010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 69113-32-2
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.14
MDL Number MFCD00216571,MFCD02730126
InChI Key OSUPWUQRPLIJKX-UHFFFAOYSA-N
Synonym 4-fluoro-n-hydroxybenzimidamide, 4-fluoro-n'-hydroxybenzimidamide, z-4-fluoro-n'-hydroxybenzenecarboximidamide, 4-fluorophenyl hydroxyimino methylamine, benzenecarboximidamide,4-fluoro-n-hydroxy, p-fluorobenzamidoxime, #, zyb
PubChem CID 5960802
SMILES NC(=NO)C1=CC=C(F)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 95.2°C to 96.1°C
Color White
Elemental Analysis 53.18 to 55.91% C, 17.72 to 18.63% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
IUPAC Name 4-fluoro-N'-hydroxybenzenecarboximidamide
Formula Weight 154.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzamidoxime
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorobenzamidoxime, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.