missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroanisole, 99%

Product Code. 10376890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10376890 25 mL Glass bottle 25mL
10192190 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10376890 Supplier Thermo Scientific Supplier No. 119310250
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 459-60-9 | C7H7FO | 126.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-60-9
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00000348
InChI Key VIPWUFMFHBIKQI-UHFFFAOYSA-N
Synonym 4-fluoroanisole, p-fluoroanisole, benzene, 1-fluoro-4-methoxy, p-fluoromethoxybenzene, p-methoxyfluorobenzene, 4-fluoroanisol, p-fluorophenyl methyl ether, anisole, p-fluoro, 1-fluoro-4-methoxy-benzene, 4-methoxyfluorobenzene
PubChem CID 9987
IUPAC Name 1-fluoro-4-methoxybenzene
SMILES COC1=CC=C(C=C1)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -45°C
Color Colorless to Yellow
Density 1.1100g/mL
Boiling Point 157°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4OCH3
Refractive Index 1.4867 to 1.4887
Quantity 25 mL
Beilstein 06, I, 98
Specific Gravity 1.11
Formula Weight 126.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Fluoroanisole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoroanisole
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.