missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroanisole, 99%, Thermo Scientific Chemicals

Product Code. 11466236
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466236 100 g 100g
11456236 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466236 Supplier Thermo Scientific Alfa Aesar Supplier No. B23048.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4-Fluoroanisole is a raw material for medicine and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoroanisole is a raw material for medicine and intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-60-9
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00000348
InChI Key VIPWUFMFHBIKQI-UHFFFAOYSA-N
Synonym 4-fluoroanisole, p-fluoroanisole, benzene, 1-fluoro-4-methoxy, p-fluoromethoxybenzene, p-methoxyfluorobenzene, 4-fluoroanisol, p-fluorophenyl methyl ether, anisole, p-fluoro, 1-fluoro-4-methoxy-benzene, 4-methoxyfluorobenzene
PubChem CID 9987
IUPAC Name 1-fluoro-4-methoxybenzene
SMILES COC1=CC=C(C=C1)F

Specifications

Melting Point -45°C
Density 1.116
Boiling Point 156°C to 158°C
Flash Point 43°C (109°F)
Refractive Index 1.488
Quantity 100 g
UN Number UN1993
Beilstein 1281425
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 126.13
Percent Purity 99%
Chemical Name or Material 4-Fluoroanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.