missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-L-phenylalanine, 98+%

Product Code. 11339955
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339955 250 mg 250mg
11319955 1 g 1g
11329955 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339955 Supplier Thermo Scientific Alfa Aesar Supplier No. L19934.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1132-68-9 | C9H10FNO2 | 183.182 g/mol

4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.

Solubility
Soluble in water and 0.5M HCl (50 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-68-9
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.182
MDL Number MFCD00063064
InChI Key XWHHYOYVRVGJJY-QMMMGPOBSA-N
Synonym 4-fluoro-l-phenylalanine, p-fluoro-l-phenylalanine, s-4-fluorophenylalanine, l-4-fluorophenylalanine, h-phe 4-f-oh, s-2-amino-3-4-fluorophenyl propanoic acid, unii-e2k2vdk6kk, l-phenylalanine, 4-fluoro, s---p-fluorophenylalanine, 2s-2-amino-3-4-fluorophenyl propanoic acid
PubChem CID 716312
ChEBI CHEBI:44909
IUPAC Name (2S)-2-amino-3-(4-fluorophenyl)propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)F

Specifications

Melting Point ∽240°C (decomposition)
Beilstein 2416148
Solubility Information Soluble in water and 0.5M HCl (50mg/ml).
Quantity 250 mg
Formula Weight 183.18
Optical Rotation −25° (c=1 in Water)
Percent Purity ≥98%
Chemical Name or Material 4-Fluoro-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.