missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)phenol, 97%, Thermo Scientific™

Product Code. 11433058
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433058 1 g 1g
11443058 5 g 5g
11453058 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433058 Supplier Thermo Scientific Alfa Aesar Supplier No. A18776.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61721-07-1
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.102
MDL Number MFCD00043875
InChI Key DHPCRFYUUWAGAH-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl phenol, 2-fluoro-5-hydroxybenzotrifluoride, 3-trifluoromethyl-4-fluorophenol, phenol, 4-fluoro-3-trifluoromethyl, alpha,alpha,alpha-4-tetrafluoro-m-cresol, fhbtf, pubchem4147, acmc-209mvr, ksc495a2b
PubChem CID 601854
IUPAC Name 4-fluoro-3-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1O)C(F)(F)F)F

Specifications

Boiling Point 84°C to 86°C (15 mmHg)
Refractive Index 1.45
Quantity 1 g
UN Number UN3265
Beilstein 2098736
Formula Weight 180.1
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.