missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)benzylamine, 94%, Thermo Scientific™

Product Code. 11328325
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328325 5 g 5g
11318325 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328325 Supplier Thermo Scientific Alfa Aesar Supplier No. L10244.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67515-74-6
Molecular Formula C8H8F4N
Molecular Weight (g/mol) 194.15
MDL Number MFCD00070814
InChI Key HZDVQEUISWBXPV-UHFFFAOYSA-O
Synonym 4-fluoro-3-trifluoromethyl benzylamine, 4-fluoro-3-trifluoromethyl phenyl methanamine, 4-fluoro-3-trifluoromethyl benzyl amine, 4-fluoro-3-trifluoromethylbenzylamine, 4-fluoro-3-trifluoromethyl-benzylamine, benzenemethanamine, 4-fluoro-3-trifluoromethyl, 4-fluoro-3-trifluoromethyl phenyl methylamine, 1-4-fluoro-3-trifluoromethyl phenyl methanamine, rarechem al bw 0670
PubChem CID 522270
IUPAC Name [4-fluoro-3-(trifluoromethyl)phenyl]methanamine
SMILES [NH3+]CC1=CC=C(F)C(=C1)C(F)(F)F

Specifications

Boiling Point 101°C (3 mmHg)
Refractive Index 1.4535
Quantity 5 g
UN Number UN2735
Beilstein 3083164
Sensitivity Air Sensitive
Formula Weight 193.15
Percent Purity 94%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.