missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)benzyl alcohol, 97%, Thermo Scientific™

Product Code. 11475736
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475736 1 g 1g
11485736 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475736 Supplier Thermo Scientific Alfa Aesar Supplier No. B20814.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67515-61-1
Molecular Formula C8H6F4O
Molecular Weight (g/mol) 194.13
MDL Number MFCD00061171
InChI Key DGBDVJVPBJMVDH-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl benzyl alcohol, 4-fluoro-3-trifluoromethyl phenyl methanol, 3-trifluoromethyl-4-fluorobenzyl alcohol, 4-fluoro-3-trifluoromethyl phenyl methan-1-ol, rarechem al bd 0323, 4-fluoro-3-trifluoromethyl benzylalcohol, benzenemethanol,4-fluoro-3-trifluoromethyl, 4-fluoranyl-3-trifluoromethyl phenyl methanol
PubChem CID 144256
IUPAC Name [4-fluoro-3-(trifluoromethyl)phenyl]methanol
SMILES OCC1=CC=C(F)C(=C1)C(F)(F)F

Specifications

Boiling Point 80°C to 85°C (0.1 mmHg)
Refractive Index 1.45
Quantity 1 g
Beilstein 1960409
Formula Weight 194.13
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.