missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)benzeneboronic acid, 98%

Product Code. 15449835
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449835 5 g 5g
15439835 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449835 Supplier Thermo Scientific Alfa Aesar Supplier No. H52483.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 182344-23-6 | C7H5BF4O2 | 207.919 g/mol

4-Fluoro-3-(trifluoromethyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-3-(trifluoromethyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methabnol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182344-23-6
Molecular Formula C7H5BF4O2
Molecular Weight (g/mol) 207.919
MDL Number MFCD07363787
InChI Key GUJYFCBXDUPORN-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl phenylboronic acid, 4-fluoro-3-trifluoromethylphenylboronic acid, 4-fluoro-3-trifluoromethyl benzeneboronic acid, 5-borono-2-fluorobenzotrifluoride, 4-fluoro-3-trifluoromethyl phenyl boronic acid, 3-trifluoromethyl-4-fluoro-phenylboronic acid, boronic acid, 4-fluoro-3-trifluoromethyl phenyl, 4-fluoro-3-trifluoromethyl benzeneboronicacid
PubChem CID 17750051
IUPAC Name [4-fluoro-3-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)F)C(F)(F)F)(O)O

Specifications

Melting Point 170°C to 174°C
Quantity 5 g
Solubility Information Soluble in methabnol.
Formula Weight 207.92
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.