missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzaldehyde, 97%, Thermo Scientific™

Catalog Number 10211193
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 42564-51-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H4FNO3
Linear Formula FC6H3(NO2)CHO
MDL Number MFCD01861388
Quantity 1g
Synonym 3-nitro-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzaldehyde, pubchem22572, acmc-1ao2q, 3-nitro-4 fluorobenzaldehyde, ksc497m9r, 2-fluoro-5-formylnitrobenzene, #, 4-fluoranyl-3-nitro-benzaldehyde
InChI Key ILKWFRCNNILIJW-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C=O)[N+](=O)[O-])F
IUPAC Name 4-fluoro-3-nitrobenzaldehyde
Molecular Weight (g/mol) 169.111
PubChem CID 598129
Formula Weight 169.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-nitrobenzaldehyde
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'