missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-3'-nitroacetophenone, 98%, Thermo Scientific™

Product Code. 10088901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10088901 5 g Glass bottle 5g
10180662 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10088901 Supplier Thermo Scientific Acros Supplier No. 366010050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 400-93-1
Molecular Formula C8H6FNO3
Molecular Weight (g/mol) 183.14
MDL Number MFCD00115369
InChI Key PTCNZDJJIOLIKQ-UHFFFAOYSA-N
Synonym 4'-fluoro-3'-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethanone, 4-fluoro-3-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethan-1-one, 1-4-fluoro-3-nitro-phenyl ethanone, 4-fluoro-3'-nitroacetophenone, ethanone, 1-4-fluoro-3-nitrophenyl, 4-fluoro-3-nitroacetophneone, 3'-nitro-4'-fluoroacetophenone
PubChem CID 2734612
IUPAC Name 1-(4-fluoro-3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 46°C to 50°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 183.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4'-Fluoro-3'-nitroacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Fluoro-3'-nitroacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.