missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid, 97%

Product Code. 15497555
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497555 1 g 1g
15407565 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497555 Supplier Thermo Scientific Alfa Aesar Supplier No. H26154.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 878572-17-9 | C8H3F4NO4 | 253.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 878572-17-9
Molecular Formula C8H3F4NO4
Molecular Weight (g/mol) 253.109
MDL Number MFCD08282778
InChI Key XCZRYPKVIJEHSU-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitro-5-trifluoromethyl benzoic acid, benzoic acid,4-fluoro-3-nitro-5-trifluoromethyl
PubChem CID 24721083
IUPAC Name 4-fluoro-3-nitro-5-(trifluoromethyl)benzoic acid
SMILES C1=C(C=C(C(=C1C(F)(F)F)F)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 120°C to 124°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 253.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-nitro-5-(trifluoromethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.