missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylphenylacetic acid, 97%, Thermo Scientific™

Product Code. 15463947
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15463947

Brand: Thermo Scientific Alfa Aesar H30525.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000520-92-2
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD09832241
InChI Key VZDGZGNZGMJNPR-UHFFFAOYSA-N
Synonym 2-4-fluoro-3-methylphenyl acetic acid, 4-fluoro-3-methylphenylacetic acid, 4-fluoro-3-methylphenyl acetic acid, 4-fluoro-3-methylphenylacetic aicd
PubChem CID 39103878
IUPAC Name 2-(4-fluoro-3-methylphenyl)acetic acid
SMILES CC1=C(C=CC(=C1)CC(=O)O)F

Specifications

Melting Point 105°C to 107°C
Quantity 1 g
Formula Weight 168.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methylphenylacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.