missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylbenzoyl chloride, 97%, Thermo Scientific™

Product Code. 11420787
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420787 1 g 1g
11430787 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420787 Supplier Thermo Scientific Alfa Aesar Supplier No. B23188.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-84-5
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.583
MDL Number MFCD01631426
InChI Key FMFLKTDRPXYUGP-UHFFFAOYSA-N
Synonym 4-fluoro-3-methylbenzoylchloride, 4-fluoro-m-toluoyl chloride, 4-fluoro-3-methyl-benzoyl chloride, benzoyl chloride, 4-fluoro-3-methyl, acmc-1arwr, 5-chlorocarbonyl-2-fluorotoluene, 4-fluoranyl-3-methyl-benzoyl chloride, 4-fluoro-3-methyl benzoyl chloride, 4-fluoro-3-methyl-benzoic acid chloride, 4-fluoro-3-methylbenzoyl chloride
PubChem CID 2774589
IUPAC Name 4-fluoro-3-methylbenzoyl chloride
SMILES CC1=C(C=CC(=C1)C(=O)Cl)F

Specifications

Density 1.22
Boiling Point 110°C to 115°C (20 mmHg)
Flash Point 98°C (208°F)
Odor Pungent
Refractive Index 1.5313
Quantity 1 g
UN Number UN3265
Beilstein 1936532
Sensitivity Moisture sensitive
Formula Weight 172.59
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methylbenzoyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.