missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylbenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 15478706
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629672-19-1
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.63
MDL Number MFCD01365818
InChI Key ALEQALYEZFNWOP-UHFFFAOYSA-N
Synonym 4-fluoro-3-methyl-benzenesulfonyl chloride, 4-fluoro-3-methylbenzenesulfonylchloride, 4-fluoro-3-methylbenzene-1-sulfonyl chloride, 4-fluoro-3-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro-3-methyl, chloro 4-fluoro-3-methylphenyl sulfone, ablock ab-12-8863, 5-chlorosulphonyl-2-fluorotoluene, 4-fluro-3-methylphenyl-sulfonylchloride, 4-fluoro-3-methylphenyl-sulfonylchloride
PubChem CID 2782758
SMILES CC1=C(F)C=CC(=C1)S(Cl)(=O)=O

Specifications

Melting Point 35°C to 40°C
Flash Point >110°C (230°F)
Quantity 250 mg
UN Number UN3261
Beilstein 3254603
Sensitivity Moisture sensitive
IUPAC Name 4-fluoro-3-methylbenzenesulfonyl chloride
Formula Weight 208.63
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Fluoro-3-methylbenzenesulfonyl chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'